Learn More
Syntenin-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22399-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors.Specifica
| Syntenin-1 | |
| Polyclonal | |
| Unconjugated | |
| SDCBP | |
| MDA 9, MDA9, Scaffold protein Pbp1, SDCBP, ST1, SYCL, Syndecan binding protein 1, Syntenin-1, TACIP18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6386 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00560 | |
| SDCBP | |
| Syntenin-1 Fusion Protein Ag18078 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |