Learn More
CELLPATH SYSTEM II+ ROUND CASSETTE
Click to view available options
Materiale:
Hoppered
Loose
Threaded
Dimensione della confezione:
pezzi 1000
pezzi 2000
pezzi 3000
Descrizione
- Read rates
- Robust and secure closure ensures specimen security
- Designed to improve reagent flow
- Large color range
- Writing surface suitable for marking with a CellMark pen
Specifica
Specifica
| Colore | White |
| Materiale | Hoppered |
| Product Type | Cassette |