Learn More
T Cell Exhaustion Marker (Flow-Directly Conjugated) Antibody Pack, Novus Biologicals™
Antibody Pack
Marca:  Novus Biologicals NBP3-12863
Codice prodotto. 18308826
  • 3135.00€ / pezzi 6
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
The Human T Cell Exhaustion Marker (Flow-Directly Conjugated) Antibody Sampler Pack contains antibodies to detect expression of inhibitory receptors associated with T cell exhaustion. T cells co-expressing these inhibitory receptors are found during chronic viral infection and within the TME of many solid tumors. Anti-PD-1 is directly conjugated to PE, while anti- CTLA-4, TIGIT, TIM-3, VISTA/B7-H5/PD-1H and LAG-3 are directly conjugated to APC. This pack contains 6 full sized vials of directly conjugated primary antibody. It will allow the user to determine the co-expression of emerging checkpoint targets like LAG-3, TIM-3, TIGIT, and VISTA with the established checkpoint target, PD-1 using flow cytometry. All antibodies have been validted for use in flow cytometry applications.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
T Cell Exhaustion Marker (Flow-Directly Conjugated)
Monoclonal
See individual datasheets
See individual datasheets
RUO
Storage is content dependent.
Flow Cytometry
Unconjugated
Immune cell exhaustion markers, immune checkpoint markers, inhibitory receptors, T cell exhaustion markers
6 Vials