Learn More
T Cell Exhaustion Marker (Flow-Directly Conjugated) Antibody Pack, Novus Biologicals™
Antibody Pack
Marca: Novus Biologicals NBP3-12863
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The Human T Cell Exhaustion Marker (Flow-Directly Conjugated) Antibody Sampler Pack contains antibodies to detect expression of inhibitory receptors associated with T cell exhaustion. T cells co-expressing these inhibitory receptors are found during chronic viral infection and within the TME of many solid tumors. Anti-PD-1 is directly conjugated to PE, while anti- CTLA-4, TIGIT, TIM-3, VISTA/B7-H5/PD-1H and LAG-3 are directly conjugated to APC. This pack contains 6 full sized vials of directly conjugated primary antibody. It will allow the user to determine the co-expression of emerging checkpoint targets like LAG-3, TIM-3, TIGIT, and VISTA with the established checkpoint target, PD-1 using flow cytometry. All antibodies have been validted for use in flow cytometry applications.
Specifica
| T Cell Exhaustion Marker (Flow-Directly Conjugated) | |
| Monoclonal | |
| See individual datasheets | |
| See individual datasheets | |
| RUO | |
| Storage is content dependent. |
| Flow Cytometry | |
| Unconjugated | |
| Immune cell exhaustion markers, immune checkpoint markers, inhibitory receptors, T cell exhaustion markers | |
| 6 Vials | |
| Human |