missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAAR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA534234
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Percent identity with other species by BLAST analysis: Human (100%) Chimpanzee (95%) Gorilla, Orangutan, Monkey (89%) Gibbon (84%).
TA1 has been reported in human brain, dorsal root ganglion, olfactory bulb, kidney, liver, lung, pancreas, prostate, skeletal muscle, small intestine, spleen, spinal cord, and stomach. An EST for TA1 has been identified from a human stomach cancer library. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates.
Specifica
| TAAR1 | |
| Polyclonal | |
| Unconjugated | |
| Taar1 | |
| MGC126874; MGC138399; RP11-295F4.9; RTAR1; Snorf33; Ta1; Taar1; TAR1; taR-1; trace amine associated receptor 1; trace amine receptor 1; trace amine-associated receptor 1; trace-amine-associated receptor 1; TRAR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 134864 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q96RJ0 | |
| Taar1 | |
| Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human TAAR1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto