Learn More
Invitrogen™ TAAR8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA534244
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Galago, Mouse (85%).
Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. Expressed in kidney and amygdala. Not expressed in other tissues or brain regions tested.
Specifica
| TAAR8 | |
| Polyclonal | |
| Unconjugated | |
| TAAR8 | |
| G protein-coupled receptor 102; Gm1149; Gm230; GPR102; G-protein coupled receptor 102; mTaar8a; Q923; Ta10; Ta11; TA5; Ta7; TAAR8; Taar8a; Taar8b; Taar8c; Tar11; TaR-11; TAR5; TaR-5; TaR-8; TaR-8a; trace amine associated receptor 8; trace amine receptor 11; Trace amine receptor 5; trace amine receptor 8; trace amine receptor 8a; trace amine-associated receptor 8; Trace amine-associated receptor 8a; trace-amine-associated receptor 8a; Trar11; Trar5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83551 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q969N4 | |
| TAAR8 | |
| Synthetic 13 amino acid peptide from N-terminal extracellular domain of human TAAR8. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |