Learn More
IBA Lifesciences TACS Column Adapter
Codice prodotto. p-7237247 Sfoglia Tutto IBA Lifesciences Prodotti
Click to view available options
Tipo di colonna:
0.3 mL
1 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 17277590
Marca: IBA Lifesciences 63333001
Adapter for Strep-Tactin™ TACS Agarose Columns that fits onto standard tubes
  • The adapters allow fitting of Strep-Tactin™ TACS Agarose columns onto standard tubes.
  • Column adapters are available in two sizes: For 3 ml columns (containing 0.3 ml Strep-Tactin™ TACS Agarose) to fit them onto 15 ml tubes and for 10 ml columns (containing 1 ml Strep-Tactin™ TACS Agarose) to fit them onto 50 ml tubes. 
Specifica
Tipo di colonna 0.3 mL