Learn More
Corning™ Tank Liners
Codice prodotto. 17213594 Sfoglia Tutto Corning Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
200 L
Diametro (sistema metrico):
55.88 cm
57.15 cm
Product Type:
Gusseted Tank Liner
Non-Gusseted Tank Liner
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Diametro (sistema metrico) Product Type unitSize
17213594 200 L 55.88 cm Gusseted Tank Liner Pezzo
17203594 200 L 57.15 cm Non-Gusseted Tank Liner Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 17213594 Fornitore Corning™ N. del fornitore 9130035
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
  • Made from low-density polyethylene (LDPE)
  • Eliminate costs associated with cleaning and validation of tanks
  • Decrease labor costs and increase turnaround time
  • Eliminate risks associated with cross contamination
  • Help extend life of reuseable tanks, resulting in reduced capital expenditures
  • Gamma irradiated, individually packaged
  • Choice of gusseted and non-gusseted available
  • Available in many sizes
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 200 L
Materiale LDPE
Capacità (sistema imperiale) 0.0528 gal.
Profondità (sistema imperiale) 36 in.
Profondità (sistema metrico) 91.44 cm
Diametro (sistema imperiale) 22 in.
Diametro (sistema metrico) 55.88 cm
Forma Cylindrical
Size 22 in. x 36 in. (55.9 cm x 91.4 cm)
Product Type Gusseted Tank Liner