Learn More
Corning™ Tank Liners
Codice prodotto. p-4668285 Sfoglia Tutto Corning Prodotti
Click to view available options
Capacità (sistema metrico):
200 L
Diametro (sistema metrico):
55.88 cm
57.15 cm
Product Type:
Gusseted Tank Liner
Non-Gusseted Tank Liner
Dimensione della confezione:
Pezzo
Codice prodotto. 17203594
Marca: Corning™ 9130030
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
  • Made from low-density polyethylene (LDPE)
  • Eliminate costs associated with cleaning and validation of tanks
  • Decrease labor costs and increase turnaround time
  • Eliminate risks associated with cross contamination
  • Help extend life of reuseable tanks, resulting in reduced capital expenditures
  • Gamma irradiated, individually packaged
  • Choice of gusseted and non-gusseted available
  • Available in many sizes
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 200 L
Materiale LDPE
Capacità (sistema imperiale) 0.0528 gal.
Profondità (sistema imperiale) 36 in.
Profondità (sistema metrico) 91.44 cm
Diametro (sistema imperiale) 22.5 in.
Diametro (sistema metrico) 57.15 cm
Forma Cylindrical
Size 22.5 in. x 36 in. (57.2 cm x 91.4 cm)
Product Type Non-Gusseted Tank Liner