Learn More
Invitrogen™ Tau Monoclonal Antibody (TAU-5)
GREENER_CHOICE
Mouse Monoclonal Antibody
Marca:  Invitrogen™ MA512808
Codice prodotto. 11304723
  • 550.00€ / 500µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
MA5-12808 targets TAU in immunofluorescence, immunoprecipitation, and Western blot applications and shows reactivity with Bovine, Human, mouse, Ovine, and Rat samples. The MA5-12808 immunogen is purified bovine microtubule-associated proteins.
Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Tau
Monoclonal
0.5 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
P10636, P10637, P19332, P29172
MAPT
Purified bovine microtubule-associated proteins.
100 μg
Primary
Bovine, Human, Mouse, Sheep, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
TAU-5
Unconjugated
MAPT
AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287)
Mouse
Protein A
RUO
101121024, 17762, 281296, 29477, 4137
-20°C
Liquid
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni