missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5105737
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Antibody detects endogenous levels of total TBC1D1.
The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.
Specifica
| TBC1D1 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D1 | |
| 1110062G02Rik; AI385682; AW555803; Kiaa1108; LYN; Lyncein; mKIAA1108; TBC; Tbc1; TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1; TBC1 domain family member 1; TBC1 domain family, member 1; Tbc1d1; tre-2/USP6, BUB2, cdc16) domain family, member 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23216, 57915 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q60949, Q86TI0 | |
| TBC1D1 | |
| A synthesized peptide derived from mouse TBC1D1(Accession Q60949), corresponding to amino acid residues P218-L238. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto