Learn More
TCF4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22337-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TCF4 is transcription factor 4, a basic helix-turn-helix transcription factor. The encoded protein recognizes an Ephrussi-box (′E-box′) binding site (′CANNTG′) - a motif first identified in immunoglobulin enhancers. This gene is expressed predomitly in pre-B-cells, although it is found in other tissues as well. Defects in this gene are a cause of Pitt-Hopkins syndrome.Specifica
| TCF4 | |
| Polyclonal | |
| Unconjugated | |
| TCF4 | |
| bHLHb19, E2 2, ITF 2, ITF2, PTHS, SEF 2, SEF2, SEF2 1, SEF2 1A, SEF2 1B, SL3 3 enhancer factor 2, TCF 4, TCF4, transcription factor 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21413, 6925 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15884, Q60722 | |
| TCF4 | |
| TCF4 Fusion Protein Ag17813 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |