missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCHH Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA555828
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: QEKSRREEQE LWQEEEQKRR QERERKLREE HIRRQQKEEQ RHRQVGEIKS QEGKGHGRLL EPGTHQFASV PVRSSPLYEY Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
This protein is an intermediate filament-associated protein that associates in regular arrays with keratin intermediate filaments (KIF) of the inner root sheath cells of the hair follicle and the granular layer of the epidermis. It later becomes cross-linked to KIF by isodipeptide bonds. It may serve as scaffold protein, together with involucrin, in the organization of the cell envelope or even anchor the cell envelope to the KIF network.
Specifica
| TCHH | |
| Polyclonal | |
| Unconjugated | |
| TCHH | |
| AHF; AI597080; hair follicle protein AHF; Tchh; THH; THL; TRHY; Trichohyalin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7062 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q07283 | |
| TCHH | |
| Recombinant protein corresponding to Human TCHH. Recombinant protein control fragment (Product #RP-94336). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto