Learn More
TCR Beta Armenian Hamster anti-Mouse, APC, Clone: H57-597, Proteintech
Armenian Hamster Monoclonal Antibody
Marca: Proteintech APC-65106-100UG
Descrizione
The TCR Beta antibody from Proteintech is a armenian hamster monoclonal antibody generated with affinity purified tcr from mouse do-11.10 cells of mouse TCR Beta. This antibody recognizes mouse antigen. The TCR Beta antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifica
| TCR Beta | |
| Monoclonal | |
| 0.2 mg/mL | |
| Armenian Hamster | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| H57-597 | |
| APC | |
| 100 μg | |
| 21577 | |
| IgG |