missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Antibody (GL-3), PE/Cy7, Novus Biologicals™
Hamster Monoclonal Antibody
Marca: Novus Biologicals FAB7297PECY7
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry
Specifica
| TCR gamma/delta | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| GL-3 | |
| Flow Cytometry | |
| T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
| Mouse intraepithelial lymphocytes | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Stem Cells | |
| 6965 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering