missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEAD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13120-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This is a rabbit polyclonal anti-TEAD3 antibody raised against part of C-terminal chain of human TEAD3.
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predomitly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon.Specifica
| TEAD3 | |
| Polyclonal | |
| Unconjugated | |
| TEAD3 | |
| DTEF 1, ETFR 1, TEA domain family member 3, TEAD 3, TEAD3, TEAD5, TEF 5, TEF5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21678, 7005 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P70210, Q99594 | |
| TEAD3 | |
| TEAD3 Fusion Protein Ag3875 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto