Learn More
Cole-Parmer™ Essentials Cell Culturing Flask for BS-200 Series Cell Culture Stirring Systems
Click to view available options
Capacità (sistema metrico):
125 mL
250 mL
500 mL
1000 mL
Dimensione della confezione:
Pezzo
Descrizione
- Completely sealed vessel is ideal for pathogenic materials
Culture vessels come calibrated and have two side ports and a central filling port. All culture vessel parts can be autoclaved at 121°C and 15 psi for 30 minutes. Silicone-encapsulated stir rods reduce the possibility of cells attaching to and growing on the surface.
Specifica
Specifica
| Capacità (sistema metrico) | 1000 mL |
| Materiale | Borosilicate Glass |
| Altezza (sistema imperiale) | 10.4 in. |
| Altezza (sistema metrico) | 263 mm |
| Autoclavabile | Autoclavable |
| Diametro (sistema metrico) esterno | 140 mm |