Learn More
Cole-Parmer™ Essentials Cell Culturing Flask for BS-200 Series Cell Culture Stirring Systems
Codice prodotto. 11855395
Click to view available options
Capacità (sistema metrico):
125 mL
250 mL
500 mL
1000 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 11855395
Marca: Cole-Parmer™ Essentials 2183734
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
This Cell Culturing Flask is a complete culture vessel—flask, stir rod, and caps—to use with biological stirrers.
  • Completely sealed vessel is ideal for pathogenic materials
Culture vessels come calibrated and have two side ports and a central filling port. All culture vessel parts can be autoclaved at 121°C and 15 psi for 30 minutes. Silicone-encapsulated stir rods reduce the possibility of cells attaching to and growing on the surface.
Specifica
Capacità (sistema metrico) 250 mL
Materiale Borosilicate Glass
Altezza (sistema imperiale) 6.7 in.
Altezza (sistema metrico) 170 mm
Autoclavabile Autoclavable
Diametro (sistema metrico) esterno 80 mm