missing translation for 'onlineSavingsMsg'
Learn More

Tedizolid, MedChemExpress

Codice prodotto. 16466208
Encompass_Preferred
Click to view available options
Quantità:
5 mg
10 mg
50 mg
Dimensione della confezione:
50mg
5mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16466208

Marca: Medchem Express HY14855

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress Tedizolid (TR 700; Torezolid; DA-7157) is a novel oxazolidinone, acting through inhibition of bacterial protein synthesis by binding to 23S ribosomal RNA (rRNA) of the 50S subunit of the ribosome.

Tedizolid (TR 700

Specifica

Materiale o nome chimico Tedizolid
CAS 856866-72-3
Colore Off-White
Note sul grado di purezza Research
Formula molecolare C17H15FN6O3
Quantità 10 mg
Sinonimo TR 700 Torezolid DA-7157
Informazioni di solubilità DMSO : 10 mg/mL (27.00 mM; Need ultrasonic)
SMILES O=C1O[C@@H](CO)CN1C2=CC=C(C3=CC=C(C4=NN(C)N=N4)N=C3)C(F)=C2
Molecular Weight (g/mol) 370.34
Peso formulazione 370.34
Percent Purity 99.19%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) COVID-19-immunoregulation
Vedi altri risultati Mostra meno risultati

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.