Learn More
Aalborg™ Model L Flowmeter
Click to view available options
Tipo:
Flowmeter
Dimensione della confezione:
Pezzo
Descrizione
- Chemically inert wetted components constructed from PFA PTFE and PCTE
- Non-fluid contacting structurally rigid frame constructed from polypropylene
- Overlapping flow ranges are available for water from 5 mLPM (0.00132 GPM) to 45 LPM (12 GPM).
- Individually leak tested
Specifica
Specifica
| Tipo | Flowmeter |
| Accuracy | ±0.5% |
| Valori nominali pressione | 6.7 bar |
| Collegamenti | 3/4 in. NPT (F) |
| Bilancia | 0 to 10 markings |
| Temperatura (sistema metrico) | 121°C |
| Valvola | Valve at outlet |
| Portata flusso d'acqua | 3000 mL/min. |