Learn More
Thermo Scientific™ Teledyne CETAC Autosamplers
Codice prodotto. p-7230077 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MS
Modello:
CETAC ASX-560 Autosampler
Dimensione della confezione:
Pezzo
Codice prodotto. 17875372
Marca: Thermo Scientific™ BRE0003260
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Increase sample throughput with advanced Teledyne CETAC Autosamplers, compatible with Thermo Scientific™ high-performance trace elemental analyzers.
Increase sample throughput in your laboratory with Teledyne CETAC Autosamplers. This selection of advanced autosamplers, compatible with Thermo Scientific™ high performance trace elemental analyzers, enables operators to take on other tasks, while intelligent, automated sample introduction accelerates workflows and eliminates manual errors.
Teledyne CETAC ASX-280 Autosampler
Accommodates up to 180 sample capacity for low-moderate sample throughput, compatible with:
  • Thermo Scientific™ iCE™ 3000 Series AAS systems
  • Thermo Scientific™ iCAP™ 7000 Series ICP-OES systems
  • Thermo Scientific™ iCAP™ PRO ICP-OES systems
  • Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS systems
  • Thermo Scientific™ Element™ Series HR-ICP-MS systems
Teledyne CETAC ASX-560 Autosampler
Accommodates up to 360 samples for moderate-high sample throughput, compatible with:
  • iCE 3000 Series AAS systems
  • iCAP 7000 Series ICP-OES systems
  • iCAP PRO ICP-OES systems
  • iCAP Q/Qnova Series ICP-MS systems
  • Element Series HR-ICP-MS systems
Teledyne CETAC XLR-860 Autosampler
Enables the unattended analysis of up to 720 samples, for laboratories looking to push the boundaries of unattended analysis, compatible with:
  • iCAP 7000 Series ICP-OES systems
  • iCAP PRO ICP-OES systems
  • iCAP Q/Qnova Series ICP-MS systems
Teledyne CETAC ASX-7400 Autosampler
Incorporates automated liquid stirring to ensure sample homogeneity, for samples containing particulates in suspension such as wear metals applications, compatible with:
  • iCAP 7000 Series ICP-OES systems
  • iCAP PRO ICP-OES systems
Specifica
Da utilizzare con (apparecchiatura) iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MS
Tipo Autosampler
Modello CETAC ASX-560 Autosampler