Learn More
Thermo Scientific™ Teledyne CETAC Autosamplers
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Increase sample throughput in your laboratory with Teledyne CETAC Autosamplers. This selection of advanced autosamplers, compatible with Thermo Scientific™ high performance trace elemental analyzers, enables operators to take on other tasks, while intelligent, automated sample introduction accelerates workflows and eliminates manual errors.
Accommodates up to 180 sample capacity for low-moderate sample throughput, compatible with:
- Thermo Scientific™ iCE™ 3000 Series AAS systems
- Thermo Scientific™ iCAP™ 7000 Series ICP-OES systems
- Thermo Scientific™ iCAP™ PRO ICP-OES systems
- Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS systems
- Thermo Scientific™ Element™ Series HR-ICP-MS systems
Accommodates up to 360 samples for moderate-high sample throughput, compatible with:
- iCE 3000 Series AAS systems
- iCAP 7000 Series ICP-OES systems
- iCAP PRO ICP-OES systems
- iCAP Q/Qnova Series ICP-MS systems
- Element Series HR-ICP-MS systems
Enables the unattended analysis of up to 720 samples, for laboratories looking to push the boundaries of unattended analysis, compatible with:
- iCAP 7000 Series ICP-OES systems
- iCAP PRO ICP-OES systems
- iCAP Q/Qnova Series ICP-MS systems
Incorporates automated liquid stirring to ensure sample homogeneity, for samples containing particulates in suspension such as wear metals applications, compatible with:
- iCAP 7000 Series ICP-OES systems
- iCAP PRO ICP-OES systems
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MS |
| Tipo | Autosampler |
| Modello | CETAC ASX-560 Autosampler |