Learn More
Sampling Systems™ TeleSwab™ Telescopic Swabbing Tool Complete Kits
Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Stile:
Kit 1
Kit 2
Include:
Angle Adapter, Swab Holder
Template Holder, Swabbing Template
Dimensione della confezione:
1 set
Descrizione
- Ideal for environmental swabbing and for use with large items of processing equipment
- Extendable handle that can be extended up to 3m
- Adjustable adapter for angling swabs to meet the contours of the item being sampled
- PTFE template tool allows a fixed area to be swabbed
- Two different complete kits are available
- Kit 1 contains one telescopic pole, one angle adapter and one swab holder
- Kit 2 contains one telescopic pole, one template holder and one swabbing template with a hole cut to customer’s requirements
Specifica
Specifica
| Product Type | Swabbing Kit |
| Include | Template Holder, Swabbing Template |
| Tipo di maniglia | Telescoping |
| Stile | Kit 2 |
| Da utilizzare con (applicazione) | Environmental swabbing |
| Lunghezza (sistema metrico) maniglia | Extendable to 3 m |