Learn More
Retsch™ Test Sieves with 1.00 to 10.00 mm Mesh Size
Codice prodotto. p-7161048 Sfoglia Tutto Retsch Prodotti
Click to view available options
Dimensione del poro:
1 mm
1.12 mm
1.18 mm
1.25 mm
1.40 mm
1.60 mm
1.70 mm
1.80 mm
2.00 mm
2.24 mm
2.36 mm
2.50 mm
2.80 mm
3.15 mm
3.35 mm
3.55 mm
4.00 mm
4.50 mm
4.75 mm
5.00 mm
5.60 mm
6.30 mm
6.70 mm
8.00 mm
9.00 mm
9.50 mm
10.00 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15598841
Marca: Retsch™ 60.131.001000
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Retsch™ Test Sieves consist of a solid stainless steel sieve frame with high stability, resulting in reliable sieving results.
  • High quality joining technology guarantees permanently taut wire mesh
  • One-piece cast sieve frame and seamless mesh joint prevent cross contamination
  • Stainless steel sieve frame with high form stability
  • High degree of corrosion resistance and easy cleaning thanks to high-alloy stainless steel
  • Excellent product quality due to extensive optical inspection
  • Maximum stability and optimum sealing when used in sieve stacks thanks to the o-ring which is placed in the recess designed for this purpose
  • Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifica
Dimensione del poro 1 mm
Tipo Test Sieve
Certificazione/Conformità DIN ISO 3310-1
Materiale Stainless Steel
Include Calibration Certificate
Altezza (sistema metrico) 50 mm
Diametro (sistema metrico) esterno 200 mm