Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Codice prodotto. p-9684005 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Colore:
Blue
Green
Orange
Red
White
Capacità:
Tubes (1.5 mL)
Tubes (10 to 13 mm)
Tubes (14 to 16 mm), Centrifuge Tubes (15 mL)
Tubes (17 to 20 mm)
Tubes (21 to 25 mm)
Tubes (26 to 30 mm)
Tubes (26 to 30 mm), Centrifuge Tubes (50 mL)
Matrice:
3 x 3
3 x 8 array
4 x 10
4 x 4
4 x 5
4 x 6
6 x 12
6 x 6
8 x 12
Dimensione della confezione:
Pezzo
Codice prodotto. 11719518
Marca: Thermo Scientific™ 30191
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
TRUSTED_SUSTAINABILITY
Specifica
Product Type Test Tube Rack
Descrizione Blue; 1.5 mL; 4 x 6 array
Matrice 4 x 6
Colore Blue
Capacità Tubes (1.5 mL)
Posizioni 24
Da utilizzare con (apparecchiatura) MaxQ™ Shaker Platform