Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint with Graduation
Codice prodotto. p-7222585
Click to view available options
Capacità (sistema metrico):
20 mL
25 mL
Dimensione della confezione:
pezzi 10
Codice prodotto. 10018289
Marca: Lenz Laborglasintrumente 05706003
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With round bottom
  • With NS socket
  • With PE stopper
  • With scale in high-contrast enamel
  • Made of DURAN™ tubing
Specifica
Capacità (sistema metrico) 25 mL
Materiale Borosilicate Glass (3.3)