Learn More
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Thin Wall
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Capacità (sistema metrico):
6 mL
10 mL
15 mL
19 mL
23 mL
25 mL
Lunghezza (sistema metrico):
75 mm
100 mm
125 mm
150 mm
160 mm
Diametro (sistema metrico):
12 mm
13 mm
16 mm
Dimensione della confezione:
pezzi 1000
Descrizione
- Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Manufactured from soda lime glass
- Thick-walled and therefore mechanically very resistant
- Available with beaded rim or straight rim
- With round bottom
Specifica
Specifica
| Capacità (sistema metrico) | 25 mL |
| Lunghezza (sistema metrico) | 160 mm |
| Materiale | Vetro sodico calcico |
| Forma | Cylindrical |
| Sterilità | Non-sterile |
| Autoclavabile | Not Autoclavable |
| Graduated | No |
| Diametro (sistema metrico) | 16 mm |
| Tipo di chiusura | Not Included |
| Product Type | Test Tube |
| Vedi altri risultati |