Learn More
Texwipe™ Replacement Covers for AlphaMop™ Cleanroom Mops
Codice prodotto. 17804085 Sfoglia Tutto Texwipe Prodotti
Click to view available options
Autoclavabile:
Autoclavable
Dimensione della confezione:
1 set
Codice prodotto. 17804085
Marca: Texwipe™ STX7118
Includes
Lot-specific certificates of compliance and irradiation
Suitable for requirements of Class 3-5 environments
  • Made of pre-sterilized double-knit AlphaWipe™ polyester
  • Gamma irradiated; covers and pads double-bagged together for sterlity assurance
  • For use in controlled, aseptic and sterile environments
  • Mopping walls, floors and ceilings
  • Applying and removing cleaning and disinfecting solutions
  • Extremely low levels of releasable fibers
  • Featured fitting compatibility with lat-padded head mop easily changeable, laundered polyester
  • Completely independent quality control audits for sterility assurance
  • Easy-to-read lot code and expiration date on each bag
  • Eliminates preparation, documentation and validation of autoclaving covers and pads
  • Conforms to textured floors and other surfaces
Compliance
Sterility Assurance Level (SAL) of 106 in accordance with ANSI™/AAMI™/ISO™11137