Learn More
Texwipe™ Thermaseal™ 70 Wipers
High absorbency, abrasion resistant
Marca: Texwipe™ TX2079
Dettagli aggiuntivi : Peso : 1.25640kg
Descrizione
- Produced, packed in ISO Class 4 cleanroom
- Hot cut
- Thermally sealed edges
- Continuous-filament
- Double-knit
- Diamensions: 23 × 23cm (9 × 9 in.)