missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFB2M Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA540897
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Peptide sequence: ECNPGPGILT QALLEAGAKV VALESDKTFI PHLESLGKNL DGKLRVIHCD Sequence homology: Horse: 100%; Human: 100%; Rabbit: 83%; Rat: 85%.
S-adenosyl-L-methionine-dependent methyltransferase which specifically dimethylates mitochondrial 12S rRNA at the conserved stem loop. Also required for basal transcription of mitochondrial DNA, probably via its interaction with POLRMT and TFAM. Stimulates transcription independently of the methyltransferase activity. Compared to TFB1M, it activates transcription of mitochondrial DNA more efficiently, while it has less methyltransferase activity.
Specifica
| TFB2M | |
| Polyclonal | |
| Unconjugated | |
| Tfb2m | |
| dimethyladenosine transferase 2, mitochondrial; FLJ22661; FLJ23182; HCV NS5A-transactivated protein 5; Hepatitis C virus NS5A-transactivated protein 5; Hkp1; h-mtTFB; h-mtTFB2; house-keeping protein 1; hTFB2M; mitochondrial 12S rRNA dimethylase 2; Mitochondrial transcription factor B2; mTFB2M; mtTFB2; NS5ATP5; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 2; TFB2M; transcription factor B2, mitochondrial | |
| Rabbit | |
| Protein A | |
| RUO | |
| 64216 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H5Q4 | |
| Tfb2m | |
| Synthetic peptide directed towards the N-terminal of human TFB2M (aa 101-150). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto