Learn More
Novus Biologicals™ TFPI Antibody Pair [HRP]
Codice prodotto. 18218107 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
15 Plates
5 Plates
Dimensione della confezione:
pezzi 15
pezzi 5
Codice prodotto. 18218107
Marca: Novus Biologicals™ NBP27953115PLATES
Rabbit Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse TFPI.
TRUSTED_SUSTAINABILITY
Specifica
Intervallo saggio 24 pg/mL
Assay Sensitivity 23.44-1500 pg/mL
Saggio Sandwich ELISA
Coniugato HRP
Product Type Antibody Pairs
Specificità This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse TFPI.
Target Species Mouse
Da utilizzare con (applicazione) Sandwich ELISA
Alias gene convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
ID gene (immissione) 7035
Simbolo del gene TFPI
Specie ospite Rabbit
Contenuto kit
  • Detection Antibody: 0.25 mg/mL rabbit anti-TFPI monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 0.4 mg/mL of rabbit anti-mouse TFPI monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 38 ng of recombinant mouse TFPI.
  • Quantità 15 Plates
    Status giuridico RUO
    Requisiti di stoccaggio Storage is content dependent.
    Bersaglio TFPI
    Tecnica 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1.5 ng/mL is recommended.