missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TfR (Transferrin R) Antibody (8D3), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NB100-64979PCP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Mouse samples. It is validated for Flow Cytometry.
Specifica
| TfR (Transferrin R) | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| TFRC | |
| Mouse transformed endothelioma cell line t.end1. | |
| 0.25 mL | |
| Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 7037 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 8D3 | |
| Flow Cytometry | |
| CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64979 recognizes native, soluble and denatured forms of murine CD71. The antibody has been used as a BBB transporter vector in mice and is suitable for studying CD71 expression, and iron uptake into different tissues, in the mouse. | |
| Store at 4C in the dark. | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto