missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TfR (Transferrin R) Antibody (OX26) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 16 publications
Marca: Novus Biologicals NB200-585-0.1MG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.
Specifica
| TfR (Transferrin R) | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular), CyTOF-ready | |
| CD71 antigen, CD71TRFR, OX26 monoclonal, OX26 transferrin receptor, p90, T9, TfR, TfR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Labels dividing cells of various types but also binds to a number of non-dividing normal tissues and endothelium of brain capillaries. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OX26 | |
| Unconjugated | |
| Q99376 | |
| TFRC | |
| PHA activated rat lymphocytes. | |
| 0.1 mg | |
| Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 7037 | |
| Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto