Learn More
Abeba™ The Original 5080 Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- White/mint
- Slip resistance outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Leather top layer
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, FO SRC/li |
| Colore | White |
| Materiale | Leather |
| Size | 35 |
| Caratteristiche suola | Slip-resistant |
| Stile | 5080 |
| Resistente a | Animal fats, oil and fuel |