Learn More
Abeba™ The Original 7021 Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB FO SRC
- Smooth leather white
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB FO SRC |
| Colore | White |
| Materiale | Leather |
| Size | 46 |
| Caratteristiche suola | Slip-resistant |
| Stile | 7021 |
| Resistente a | Animal fats, oil and fuel |