Learn More
Abeba™ The Original 7201 Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 paio
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White/black
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
- heel cushion
Specifications
Specifications
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, FO SRC |
| Colore | White |
| Materiale | Leather |
| Size | 35 |
| Caratteristiche suola | Slip-resistant |
| Stile | 7201 |
| Resistente a | Animal fats, oil and fuel |