Learn More
Thermo Scientific™ TheraPure™ Modifying Enzymes
Codice prodotto. 18416728 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
10 U, 10 μL
10 U, 100 μL
10,000 U, 250 μL
5000 U, 100 μL
Concentrazione:
0.1 U/μL
1 U/μL
40 U/μL
50 U/μL
Product Type:
DNase I, RNase-free
Pyrophosphatase, Inorganic
RNase Inhibitor
Dimensione della confezione:
100µL
10µL
250µL
Codice prodotto. 18416728
Marca: Thermo Scientific™ EF0221T1
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.
The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.
Features
  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product
The exceptional quality of TheraPure products is supported by:
  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement
TRUSTED_SUSTAINABILITY
Specifica
Concentrazione 1 U/μL
Da utilizzare con (applicazione) In vitro Transcription
Applicazione validata RNA synthesis
Enzyme Pyrophosphatase, inorganic
Sensitive to Heat Inactivation No
Quantità 10 U, 10 μL
Product Type Pyrophosphatase, Inorganic
Peso molecolare 32 kDa
Product Line TheraPure
Inibitori Imidodiphosphate, α ω-glycol disphosphates, methanedial diphosphate, 1,2-ethanedial diphosphate
Forma Liquid
Esclusivamente per scopi di ricerca.