Learn More
Thermo Scientific™ Thermo Scientific Capit-All Flex Matrix/Nunc Cap Driver Cassette
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Formato decapsulazione:
96-Channel
Dimensione della confezione:
Pezzo
Descrizione
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, It is recommended to be replaced after 10,000 decapping cycles.
- Cap driver cassette compatible with Thermo Scientific Capit-All Flex Automated Decapper (P/N 4120-FLEX)
- Tube compatibility: Matrix and Nunc cryotubes in 96 and 48 formats
- Replace after 10,000 cycles
Specifica
Specifica
| Provette per conservazione compatibili | Matrix Screw Top Tubes (0.2, 0.5, 1 mL), Nunc Cryobank Tubes (0.5, 1, 2 mL), Nunc Universal Tubes (1.8, 2 mL) |
| Formato decapsulazione | 96-Channel |
| Funzionamento | Touchscreen User Interface |
| Tipo | Decapping Head |