Learn More
Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for GC Instruments
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Choose Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for accurate, reproducible GC sample introduction using an affixed needle and a plunger that creates a tight seal.
Get durability, clarity and accuracy in gas chromatography (GC) sample introduction for confidence in analytical results, time-after-time. Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes offer an affixed needle and a plunger that creates a tight seal with the barrel. Choose from a variety of sizes, types and gas chromatography autosampler compatibilities. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifica
Specifica
| Calibro dell'ago | 23 to 26 G |
| Lunghezza (sistema metrico) | 42 mm |
| Product Type | Gas Tight GC Autosampler Syringe, Fixed Needle |
| Quantità | 1/Ea. |
| Volume (sistema metrico) | 10 μL |
| Product Line | Gastight |
| Lunghezza (sistema metrico) ago | 42 mm |
| Stile del punto dell'ago | Cone |
| Da utilizzare con (apparecchiatura) | Agilent GC Instruments |
| Tipo di collegamento | Fixed |