Learn More
Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for GC Instruments
Codice prodotto. p-4467126 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Volume (sistema metrico):
10 μL
25 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
Calibro dell'ago:
23 G
23 to 26 G
26 G
26s G
Lunghezza (sistema metrico):
42 mm
50 mm
57 mm
85 mm
Stile del punto dell'ago:
Cone
Side Hole
Dimensione della confezione:
Pezzo
pezzi 6
Codice prodotto. 10515995
Marca: Thermo Scientific™ 365D2976
Choose Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for accurate, reproducible GC sample introduction using an affixed needle and a plunger that creates a tight seal.
Get durability, clarity and accuracy in gas chromatography (GC) sample introduction for confidence in analytical results, time-after-time. Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes offer an affixed needle and a plunger that creates a tight seal with the barrel. Choose from a variety of sizes, types and gas chromatography autosampler compatibilities. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
Durability
The enhanced design of the syringes delivers:
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
Clarity
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
Accuracy
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover:
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifica
Materiale Glass (Barrel), PTFE (Insert)
Calibro dell'ago 26 G
Lunghezza (sistema metrico) 50 mm
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Quantità 6/Pk.
Volume (sistema metrico) 10 μL
Product Line Gastight
Lunghezza (sistema metrico) ago 50 mm
Stile del punto dell'ago Cone
Da utilizzare con (apparecchiatura) CTC GC Instruments, AI/AS 1610 Autosamplers
Tipo di collegamento Fixed