Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer COD Vial Cell Holder
Marca: Thermo Scientific™ 4014-000
Dettagli aggiuntivi : Peso : 0.12600kg
Descrizione
COD Vial Cell Holder. Snap-in” holder can handle one COD vial up to 104mm tall and 18mm in diameter.
Specifica
Cell Holder | |
Genesys™ 20 spectrophotometer | |
One COD vial up to 104 mm tall and 18 mm dia. | |
104 mm |