Learn More
Thermo Scientific™ Provette per la raccolta a basso legame proteico
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità (sistema metrico):
1.5 mL
2 mL
Quantità:
250
2500
Dimensione della confezione:
pezzi 250
pezzi 2500
Descrizione
Thermo Scientific Pierce Low Protein Binding Microcentrifuge Tubes (1.5 or 2 mL) help minimize protein sample loss during sample manipulation and are ideal in applications where processing steps are extensive.
Pierce Low Protein Binding Microcentrifuge Tubes help minimize sample loss in mass spectrometry and immunoprecipitation/co-immunoprecipitation applications. The tubes are conveniently packaged at 250 tubes per package to minimize contamination even when purchased in bulk.
• Durable—use at speeds up to 20,000 RCF
• Compatible—suitable for IP and mass spectrometry applications
• Nuclease-free—certified RNase/DNase-free polypropylene tubes
• Durable—use at speeds up to 20,000 RCF
• Compatible—suitable for IP and mass spectrometry applications
• Nuclease-free—certified RNase/DNase-free polypropylene tubes
Specifica
Specifica
| Descrizione | Low Protein Binding Tube |
| Capacità (sistema metrico) | 1.5 mL |
| Volume (sistema metrico) | 1.5 mL |
| Linea di prodotti | Pierce |
| Quantità | 2500 |
| Product Type | Collection Tube |