Learn More
Thermo Scientific™ Nalgene™ Centrifuge Bottle Sealing Cap Assembly
GREENER_CHOICE
Codice prodotto. p-74220
Click to view available options
Include:
Blue Silicone Gasket
Red Silicone Gasket
Dimensione della confezione:
pezzi 6
Codice prodotto. 10192554
Marca: Thermo Scientific™ DS3132BLUE
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Includes
Includes polypropylene screw cap and shoulder ring, Tefzel™ ETFE plug, and Viton™ fluoroelastomer O-ring to form a leakproof† seal
Seals centrifuge bottle
  • For use with 1L centrifuge bottles
  • Autoclavable
  • Polypropylene screw closure
  • For use at recommended maximum speeds (10,000 x g or greater) without leakage, even when full
TRUSTED_SUSTAINABILITY
Specifica
Colore Blue
Include Blue Silicone Gasket
Materiale Polypropylene
Da utilizzare con (apparecchiatura) 1 L PC Nalgene Centrifuge Bottle