Learn More
Thermo Scientific™ Guarnizioni a capsula da 20 mm con setti per l’analisi dello spazio di testa National™
Click to view available options
Colore:
Ambra
Dimensione della confezione:
pezzi 100
Descrizione
20mm Crimp seals with prefitted septa for headspace vials
- Seals with pre-inserted septa are convenient and minimize contamination due to handling
- Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
- PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
- Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
Order Info
Septa and crimp seals also sold separately.
Specifica
Specifica
| Colore | Ambra |
| Materiale | Vetro di silice |
| Tipo | Setti |