Learn More
Thermo Scientific™ Graphite/Vespel Ferrule for Transfer Line
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
10/Pk
Dimensione della confezione:
pezzi 10
Descrizione
The Thermo Scientific SWAP-IT GCMS Interface provides a powerful tool for switching capillary columns without the need to vent the MS.
- Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
- Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
- The system works without the requirement for additional He-purge/make-up gas flow
- Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
- Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
Specifica
Specifica
| Tipo | Ferrule |
| Descrizione | Graphite/Vespel Ferrule; For Thermo Scientific Transfer Line |
| Quantità | 10/Pk |
| Da utilizzare con (apparecchiatura) | DSQ, DSQ II, PolarisQ, ITQ |