Learn More
Thermo Scientific™ pMCS minP-Tluc16-DD Vector for Luciferase Assays
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
The Thermo Scientific pMCS Tluc16 Vectors are multiple cloning site (MCS) vectors designed to accept a promoter sequence for study of gene regulation using the intracellular TurboLuc™16 (Tluc16) luciferase reporter.
- Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systems
- Multiple cloning sites provide versatility for transfer of regulatory elements from one vector to another
- Synthetic polyA terminator and Transcriptional Pause Site (TPS) included upstream of MCS to minimize non-specific transcriptional read-through
- Dual-destabilization (DD) technology reduces accumulation of Tluc16 luciferase mRNA and protein in cells, enhancing the responsiveness of the assay
- Optimized minimal core promoter (minP) and 5' UTR region for efficient expression of Tluc16 luciferase
- Hygromycin resistance gene for use as selection marker to generate mammalian stable cell lines
- Prepared pNF-κB, and pCRE Tluc16-DD Vectors are designed to monitor the activation of NFκB protein and cAMP-binding protein (CREB) in mammalian cells
- Prepared pEF1α variety includes a modified version of the Elongated Factor 1α (EF1α) promoter for constitutive Tluc16 luciferase expression in mammalian cells
Specifica
Specifica
| Tipo | Transcriptional Reporter Vector |
| Cloning Method | Restriction Enzyme ⁄ MCS |
| Tipo di erogazione | Transfection |
| Quantità | 10 μg |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Gene reporter | TurboLuc™16 Luciferase |
| Da utilizzare con (applicazione) | Reporter Assays |
| Promotore | minP |