Learn More
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple
Product Code. 11841280 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (English):
120 in. (probe)
60 in. (probe)
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
11841280 120 in. (probe) Each
11706624 60 in. (probe) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11841280 Supplier Cole-Parmer™ Supplier No. 0851282
Spedizione stimata: 07-01-2027
Use these probes for a variety of high-temperature applications. Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple Probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Flexible Kapton-insulated wire is ideal for checking food temperatures
  • Flexible fiberglass-Insulated wire used for high-temperature measurements
  • Custom designs or changes to this product are available upon request
  • Exposed-junction probes are not recommended for use with aqueous solutions
  • Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
Temperature (Metric) -150°C to +482°C
Type Thermocouple Probe
For Use With (Application) Used for High-Temperature Measurements
Length (English) 120 in. (probe)