Learn More
ThermoFlex™ Recirculating Chillers
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità di raffreddamento:
1400 W
Dimensione della confezione:
Pezzo
Descrizione
Optimized for the most demanding applications, Thermo Scientific™ ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain. The ThermoFlex platform was developed with customer input from concept to design. This high-performance chiller platform, with cooling capacity up to 24000w/81964 BTU, is configurable to meet application demands.
Ease of Use
- An intuitive user interface for ease of operation
- Air and water filters that can be changed while unit is in operation
- Innovative packaging for rapid installation
- Quick start guide enables trouble-free system start-up
Superior Performance
- Improved cooling capacity
- Increased reliability
- Ease of maintenance
Configurable Design
- Wide range of available cooling capacities
- Variety of available options
- Installation flexibility
Recommended for:
- Aerospace
- Analytical
- Biotech
- Chemical
- Industrial
- Medical
- Pharmaceutical
- Research
- University
Specifica
Specifica
| Temperatura ambiente | 10°C to 40°C |
| Capacità (sistema imperiale) | 1.9 gal. |
| Descrizione | 1400w; Pumping option PD1; 230V 50Hz |
| Capacità (sistema metrico) | 7.2 L |
| Certificazione/Conformità | NRTL Certified to CSA Standards; CE marked |
| Capacità di raffreddamento | 1400 W |
| Gamma di temperatura | 5°C to 40°C |
| Stabilità termica | ±° |
| Electrical Requirements | 230 V 50 Hz |
| Tensione | 230 V |
| Vedi altri risultati |