Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Codice prodotto. p-7075424
Click to view available options
Dimensione del poro:
0.4 μm
1.0 μm
3 μm
3.0 μm
8.0 μm
No. of Wells:
6
12
24
Densità poro:
0.15 x 106 pores/cm2
0.6 x 106 pores/cm2
1 x 108 pores/cm2
2 x 106 pores/cm2
Dimensione della confezione:
pezzi 24
pezzi 48
Codice prodotto. 10689094
Marca: Greiner Bio-One 657630
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
  • Clear polystyrene housing with sealed PET capillary pore membrane
  • Hanging geometry provides protection to lower compartment cell layer
  • Simplified pipetting due to self-lift geometry
  • Eccentric in-well positioning allows unequalled access to lower compartment
  • Available in 6, 12 and 24 well format
TRUSTED_SUSTAINABILITY
Specifica
Materiale Polystyrene housing with sealed PET capillary pore membrane
Densità poro 0.6 x 106 pores/cm2
Dimensione del poro 3 μm
No. of Wells 6