missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thioredoxin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA587014
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Purity is > 95% by SDS-PAGE.
Thioredoxins (Trx) are small, multi-functional proteins with oxidoreductase activity and are ubiquitous in essentially all living cells. Trx contains a redox-active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. The two cysteine residues in the conserved active centers can be oxidized to form intramolecular disulfide bonds. Reduction of the active site disulfide in oxidized Trx is catalyzed by Trx reductase with NADPH as the electron donor. The reduced Trx is a hydrogen donor for ribonucleotide reductase, the essential enzyme for DNA synthesis, and a potent general protein disulfide reductase with numerous functions in growth and redox regulations. Specific protein disulfide targets for reduction by Trx include protein disulfide-isomerase (PDI) and a number of transcription factors such as p53, NF-kB and AP-1 (T1-151). Trx is also capable of removing H2O2, particularly when it is coupled with either methionine sulfoxide reductase or several isoforms of peroxiredoxins.
Specifica
| Thioredoxin 1 | |
| Polyclonal | |
| Unconjugated | |
| TXN | |
| ADF; ATL-derived factor; AW550880; DKFZp686B1993; MGC61975; RP11-427L11.1; SASP; surface-associated sulphydryl protein; testicular tissue protein Li 199; thioredoxin; thioredoxin 1; thioredoxin delta 3; TRDX; TRX; TRX1; Txn; TXN delta 3; Txn1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22166, 7295 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P10599, P10639 | |
| TXN, Txn1 | |
| A synthetic peptide of human TXN. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto