Learn More
Euro Protection™ Thor Multirisk Pants, Navy
Codice prodotto. p-7146811
Click to view available options
Size:
Small
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
Pezzo
Codice prodotto. 15830892
Marca: Euro Protection™ 8MTHTNS
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Euro Protection™ Thor Multirisk Pants, Navy are antistatic, fire-retardant, and feature reflective band tape for visibility.
  • Fire retardant
  • Antistatic
  • Reflective band tape
  • Cotton/polyester construction
Specifica
Size Small
Product Type Multirisk Pants
Materiale Cotton
Resistente a Fire-retardant, Anti-static
Certifications/Compliance (Europe) EN ISO 11611:2007Class 1 - A1IEC 61482-2:2009 Class 1 - 4 kAEN 13034:2005+A1:2009Type PB6EN 1149-5:200N 14404:2004+A1:2010Type 2
Stile Includes reflective band tape