Learn More
SymLine™ Thread Adapter, Type 229
Sfoglia Tutto SCAT Europe Prodotti
Click to view available options
Dimensione chiusura:
S60/61, S50
Dimensione della confezione:
1 foglio
Descrizione
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifica
Specifica
| Dimensione chiusura | S60/61, S50 |
| Colore chiusura | Black |
| Descrizione | S60/61 (m) to S50 (f), excentric with FKM gasket |
| Materiale rivestimento interno | PE-HD-EL |
| Product Type | Thread Adapter |